Learn More
2,4'-Dibromoacetophenone, 98%
CAS: 99-73-0 | C8H6Br2O | 277.94 g/mol
Supplier: Thermo Scientific Chemicals 112680500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
2, 4'-Dibromoacetophenone | |
97.5 | |
107°C to 111°C | |
1415°C | |
97.5% min. (GC) | |
C8H6Br2O | |
MFCD00000200 | |
07, 285 | |
15, 1438 | |
Solubility in water: insoluble. Other solubilities: very soluble in warm alcohol,soluble in ethyl alcohol,ethyl ether,chloroform | |
C1=CC(=CC=C1C(=O)CBr)Br | |
277.94 | |
277.94 | |
Crystalline Solid |
99-73-0 | |
100.0 | |
Yellow to Beige | |
Authentic | |
Glass bottle | |
BrC6H4COCH2Br | |
50 g | |
01,77; 03,34 | |
2,4'-dibromoacetophenone, 2-bromo-1-4-bromophenyl ethanone, 4-bromophenacyl bromide, p-bromophenacyl bromide, ethanone, 2-bromo-1-4-bromophenyl, 4'-bromophenacyl bromide, p-bromophenacyl-8, alpha,p-dibromoacetophenone, alpha,4-dibromoacetophenone, 4-bromo bromoacetyl benzene | |
FKJSFKCZZIXQIP-UHFFFAOYSA-N | |
2-bromo-1-(4-bromophenyl)ethanone | |
7454 | |
98% |
Identifiants chimiques
99-73-0 | |
277.94 | |
FKJSFKCZZIXQIP-UHFFFAOYSA-N | |
7454 | |
C1=CC(=CC=C1C(=O)CBr)Br |
C8H6Br2O | |
MFCD00000200 | |
2,4'-dibromoacetophenone, 2-bromo-1-4-bromophenyl ethanone, 4-bromophenacyl bromide, p-bromophenacyl bromide, ethanone, 2-bromo-1-4-bromophenyl, 4'-bromophenacyl bromide, p-bromophenacyl-8, alpha,p-dibromoacetophenone, alpha,4-dibromoacetophenone, 4-bromo bromoacetyl benzeneAfficher plus de résultats | |
2-bromo-1-(4-bromophenyl)ethanone |
Sécurité et manipulation
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-783-6
RUO – Research Use Only