missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trimethylphenyl Isothiocyanate 98.0+%, TCI America™
Supplier: TCI America T332425G
Spécifications
| 2,4,6-Trimethylphenyl Isothiocyanate | |
| 63°C | |
| 75°C | |
| MFCD00022055 | |
| 2811 | |
| KKYMYPLVBCVDPL-UHFFFAOYSA-N | |
| 2-isothiocyanato-1,3,5-trimethylbenzene | |
| 138659 | |
| ≥98.0% (GC) |
| 6095-82-5 | |
| White-Yellow | |
| C10H11NS | |
| 25 g | |
| 2,4,6-trimethylphenyl isothiocyanate, mesityl isothiocyanate, 2,4,6-trimethylphenylisothiocyanate, 2-isothiocyanato-1,3,5-trimethyl-benzene, benzene, 2-isothiocyanato-1,3,5-trimethyl, 2,4,6-trimethylbenzenisothiocyanate, mesitylisothiocyanate, acmc-20aoen, isothiocyanic acid mesityl ester, # | |
| CC1=CC(=C(C(=C1)C)N=C=S)C | |
| 177.265 | |
| 177.27 | |
| Crystalline Powder |
Identifiants chimiques
| 6095-82-5 | |
| 177.265 | |
| KKYMYPLVBCVDPL-UHFFFAOYSA-N | |
| 138659 | |
| CC1=CC(=C(C(=C1)C)N=C=S)C |
| C10H11NS | |
| MFCD00022055 | |
| 2,4,6-trimethylphenyl isothiocyanate, mesityl isothiocyanate, 2,4,6-trimethylphenylisothiocyanate, 2-isothiocyanato-1,3,5-trimethyl-benzene, benzene, 2-isothiocyanato-1,3,5-trimethyl, 2,4,6-trimethylbenzenisothiocyanate, mesitylisothiocyanate, acmc-20aoen, isothiocyanic acid mesityl ester, # | |
| 2-isothiocyanato-1,3,5-trimethylbenzene |
Sécurité et manipulation
missing translation for 'tsca' : No