missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylphenyl Isothiocyanate 98.0+%, TCI America™

Catalog No. p-7137698
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33245G 5 g
T332425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33245G Supplier TCI America Supplier No. T33245G

Chemical Identifiers

CAS 6095-82-5
Molecular Formula C10H11NS
Molecular Weight (g/mol) 177.265
MDL Number MFCD00022055
InChI Key KKYMYPLVBCVDPL-UHFFFAOYSA-N
Synonym 2,4,6-trimethylphenyl isothiocyanate, mesityl isothiocyanate, 2,4,6-trimethylphenylisothiocyanate, 2-isothiocyanato-1,3,5-trimethyl-benzene, benzene, 2-isothiocyanato-1,3,5-trimethyl, 2,4,6-trimethylbenzenisothiocyanate, mesitylisothiocyanate, acmc-20aoen, isothiocyanic acid mesityl ester, #
PubChem CID 138659
IUPAC Name 2-isothiocyanato-1,3,5-trimethylbenzene
SMILES CC1=CC(=C(C(=C1)C)N=C=S)C

Specifications

Melting Point 63°C
Color White-Yellow
Boiling Point 75°C
Quantity 5 g
UN Number 2811
Formula Weight 177.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trimethylphenyl Isothiocyanate