missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(3,5-Dibromophenyl)-4,6-diphenyl-1,3,5-triazine 98.0+%, TCI America™
Supplier: TCI America D49021G
Spécifications
| 2-(3,5-Dibromophenyl)-4,6-diphenyl-1,3,5-triazine | |
| 246°C | |
| C21H13Br2N3 | |
| 1 g | |
| C1=CC=C(C=C1)C2=NC(=NC(=N2)C3=CC(=CC(=C3)Br)Br)C4=CC=CC=C4 | |
| 467.164 | |
| 467.16 | |
| Crystalline Powder |
| 1073062-59-5 | |
| Yellow | |
| MFCD25562933 | |
| DRQMSTXYCLCAHO-UHFFFAOYSA-N | |
| 2-(3,5-dibromophenyl)-4,6-diphenyl-1,3,5-triazine | |
| 58382615 | |
| ≥98.0% (HPLC,N) |
Identifiants chimiques
| 1073062-59-5 | |
| 467.164 | |
| DRQMSTXYCLCAHO-UHFFFAOYSA-N | |
| 2-(3,5-dibromophenyl)-4,6-diphenyl-1,3,5-triazine |
| C21H13Br2N3 | |
| MFCD25562933 | |
| 58382615 | |
| C1=CC=C(C=C1)C2=NC(=NC(=N2)C3=CC(=CC(=C3)Br)Br)C4=CC=CC=C4 |
Sécurité et manipulation
missing translation for 'tsca' : No