missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(3,5-Dibromophenyl)-4,6-diphenyl-1,3,5-triazine 98.0+%, TCI America™
$128.62 - $421.27
Chemical Identifiers
| CAS | 1073062-59-5 |
|---|---|
| Molecular Formula | C21H13Br2N3 |
| Molecular Weight (g/mol) | 467.164 |
| MDL Number | MFCD25562933 |
| InChI Key | DRQMSTXYCLCAHO-UHFFFAOYSA-N |
| PubChem CID | 58382615 |
| IUPAC Name | 2-(3,5-dibromophenyl)-4,6-diphenyl-1,3,5-triazine |
| SMILES | C1=CC=C(C=C1)C2=NC(=NC(=N2)C3=CC(=CC(=C3)Br)Br)C4=CC=CC=C4 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D4902200MG
|
TCI America
D4902200MG |
200 mg |
Each for $128.62
|
|
|||||
|
D49021G
|
TCI America
D49021G |
1 g |
Each for $421.27
|
|
|||||
Chemical Identifiers
| 1073062-59-5 | |
| 467.164 | |
| DRQMSTXYCLCAHO-UHFFFAOYSA-N | |
| 2-(3,5-dibromophenyl)-4,6-diphenyl-1,3,5-triazine |
| C21H13Br2N3 | |
| MFCD25562933 | |
| 58382615 | |
| C1=CC=C(C=C1)C2=NC(=NC(=N2)C3=CC(=CC(=C3)Br)Br)C4=CC=CC=C4 |
Specifications
| 1073062-59-5 | |
| Yellow | |
| MFCD25562933 | |
| DRQMSTXYCLCAHO-UHFFFAOYSA-N | |
| 2-(3,5-dibromophenyl)-4,6-diphenyl-1,3,5-triazine | |
| 58382615 | |
| ≥98.0% (HPLC,N) | |
| 2-(3,5-Dibromophenyl)-4,6-diphenyl-1,3,5-triazine |
| 246°C | |
| C21H13Br2N3 | |
| 200 mg | |
| C1=CC=C(C=C1)C2=NC(=NC(=N2)C3=CC(=CC(=C3)Br)Br)C4=CC=CC=C4 | |
| 467.164 | |
| 467.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : No