Learn More
1,1,1,3,3,3-Hexamethyldisilazane, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 120580025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
1,1,1,3,3,3-Hexamethyldisilazane | |
0.7600g/mL | |
20°C | |
Glass Bottle | |
1.4060 to 1.4090 | |
2.5L | |
15,4725 | |
Solubility in water: reacts. Other solubilities: reacts with methanol, soluble in acetone, benzene, ethyl ether, pentane, and perchloroethylene | |
C[Si](C)(C)N[Si](C)(C)C | |
161.4 | |
0.9 mm2/s (25°C) | |
161.4 |
999-97-3 | |
125.0°C | |
97.5% min. (GC) | |
C6H19NSi2 | |
(CH3)3SiNHSi(CH3)3 | |
0.76 | |
hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl | |
FFUAGWLWBBFQJT-UHFFFAOYSA-N | |
[dimethyl-(trimethylsilylamino)silyl]methane | |
13838 | |
CHEBI:85068 | |
98% |
Sécurité et manipulation
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 213-668-5
RUO â Research Use Only