Learn More
Axygen™ 0.5 mL Conical Screw Cap Tubes: Presterilized
Features chemical resistant O-ring
Supplier:  Axygen™ SCT050OS
 Afficher plus de versions de ce produit
Numéro de catalogue. 14-222-529
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping of restriction enzymes and other reagents.
  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
  • Available in range of colors for easy sample identification
Spécifications
Spécifications
RNase, DNase, pyrogen-free
Polypropylene
Polypropylene
O-ring
Sterile
Conical
Orange
0.5 mL
Non-looped
500 Tubes and caps/Pk.; 8 Packs/Cs.
Screw Cap
20,000 x g
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS