missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 0.5 mL Conical Screw Cap Tubes: Presterilized
Features chemical resistant O-ring
$2394.48 - $2487.22
Specifications
| Certifications/Compliance | RNase, DNase, pyrogen-free |
|---|---|
| Closure Material | Polypropylene |
| Capacity (Metric) | 0.5 mL |
| Material | Polypropylene |
| Format | Non-looped |
| Catalog Number | Mfr. No. | Closure Color | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Closure Color | Price | Quantity | |||||
|
14-222-529
|
Axygen™
SCT050OS |
Orange |
Case of 4000 Each for $2,487.22
|
|
|||||
|
50-400-780
|
Axygen™
SCT050AS |
Assorted |
Case for $2,394.48
|
|
|||||
|
14-222-561
|
Axygen™
SCT050YS |
Yellow |
Case of 4000 Each for $2,487.22
|
|
|||||
|
50-400-784
|
Axygen™
SCT050CS |
Clear |
Case for $2,416.15
|
|
|||||
|
14-222-532
|
Axygen™
SCT050RS |
Red |
Case of 4000 Each for $2,487.22
|
|
|||||
|
14-222-527
|
Axygen™
SCT050GS |
Green |
Case of 4000 Each for $2,487.22
|
|
|||||
|
14-222-521
|
Axygen™
SCT050BS |
Blue |
Case of 4000 Each for $2,487.22
|
|
|||||
Description
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping of restriction enzymes and other reagents.
- Presterilized cap tubes have non-looped caps attached with O-rings
- Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
- Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
- Available in range of colors for easy sample identification
Specifications
| RNase, DNase, pyrogen-free | |
| 0.5 mL | |
| Non-looped | |
| Sterile | |
| Conical |
| Polypropylene | |
| Polypropylene | |
| O-ring | |
| Screw Cap | |
| 20,000 x g |