UserName
Learn More About QuantStudio Real-Time PCR & Digital PCR Systems
Learn More About QuantStudio Real-Time PCR & Digital PCR Systems
Every lab is unique. That’s why you deserve a real-time PCR or digital PCR platform that fits your needs. Perhaps you’re looking for simplicity on a budget, or reliable results from limited samples. Maybe your research requires high throughput for maximum productivity, or absolute answers to take your work to the next level. Whatever you need, there’s an Applied Biosystems™ QuantStudio™ Real-Time PCR or Digital PCR System that’s just right for your research.
Visit fishersci.ca/qpcrreagents to find qPCR reagents to use with your QuantStudio System.
Filename
applied-biosystems-pcr-solutions-brochure.pdf
Size
22 MB
Format
application/pdf
Filename
finance-no-payment-flyer-americas-bn-col14678.pdf
Size
610 KB
Format
application/pdf
Filename
fisher-scientific-pcr-solutions-brochure.pdf
Size
6 MB
Format
application/pdf
Filename
applied-biosystems-genetic-analysis-in-gene-therapy-workflows.pdf
Size
1 MB
Format
application/pdf
Contact a Life Sciences Specialist
Fill out this form to get more information about which QuantStudio Real-Time PCR or Digital PCR System is right for you.
Step One: Account Information
Required Field *
Step Two: Company Information
Required Field *
State / Province
Country
Step Three: Additional Information
Required Field *
By submitting your data, you agree to our Privacy Policy and will receive emails based on your preferences.
© Thermo Fisher Scientific, Inc. All Rights Reserved.
Thermo Fisher Scientific Logo
QuantStudio 3 Real-Time PCR System
Connect to your data like never before with the Applied Biosystems™ QuantStudio™ 3 Real-Time PCR System. The small, affordable system was designed for easy use and delivers the high-performance analysis you expect.
QuantStudio 3 Real-Time PCR System
QuantStudio 5 Real-Time PCR System
With the latest advancements in touchscreen usability, the QuantStudio 5 System lets you access your data more easily than ever before and securely share results with collaborators around the world.
QuantStudio 5 Real-Time PCR System
QuantStudio 6 Pro Real-Time PCR System
Get high quality and excellent reliability with the QuantStudio 6 Pro System, which features 96- or 384-well sample capacity, five filter or color combinations, and five multiplexing targets.
QuantStudio 6 Pro Real-Time PCR System
QuantStudio 7 Pro Real-Time PCR System
The QuantStudio 7 Pro System delivers a smart workflow experience and features 96- and 384-well sample capacity, six filter or color combinations, and 21 multiplexing targets.
QuantStudio 7 Pro Real-Time PCR System
QuantStudio 6 Flex Real-Time PCR System
Prepare for the future with the QuantStudio 6 Flex System. Offering an upgradeable path to the QuantStudio 7 Flex System to support automation or Applied Biosystems™ TaqMan® Array Cards, it’s ideal for labs with multiple applications or changing throughput needs.
QuantStudio 6 Flex Real-Time PCR System
QuantStudio 7 Flex Real-Time PCR System
Get reliability, sensitivity, accuracy, and flexibility with the QuantStudio 7 Flex System. It makes moving from application to application easier than ever with additional dyes, formats, and automation potential.
QuantStudio 6 Flex Real-Time PCR System
QuantStudio 12K Flex Real-Time PCR System
Prepare for multiple types of experiments and users with one instrument: the QuantStudio 12K Flex System. With high-throughput technology, it supports maximum productivity with minimum effort.
QuantStudio 12K Flex Real-Time PCR System
QuantStudio Absolute Q Digital PCR System
The Applied Biosystems™ QuantStudio™ Absolute Q™ Digital PCR (dPCR) System is powered by proprietary microfluidic array plate (MAP) technology that lets you perform dPCR compartmentalizing, thermal cycling, and data acquisition on a single instrument. The dPCR workflow is the same as that for qPCR, which facilitates ease of use, minimizes hands on steps, and maximizes consistency.
QuantStudio Absolute Q Digital PCR System
QuantStudio™ Real-Time PCR Systems Comparison
FeatureQuantStudio™ 3QuantStudio™ 5QuantStudio™ 6 ProQuantStudio™ 7 ProQuantStudio™ 12K Flex
Block formatFixed blockFixed blockFixed block (Upgradeable to QS7)Interchangeable blockInterchangeable block
Blocks96-well (0.1 mL or 0.2 mL)96-well (0.1 mL or 0.2 mL); 384-well96-well (0.1 mL or 0.2 mL); 384-well96-well; 384-well; TaqMan™ Array Card96-well; 384-well; TaqMan™ Array Card; OpenArray™
MultiplexingUp to 4 targets96-well: up to 6 targets; 384-well: up to 5 targetsUp to 5 targetsUp to 6 targets (96- and 384-well)96-/384-well: up to 6 targets; TaqMan™ Array Card: up to 3; OpenArray™: up to 2
Automation-readyNoNoUpgradeable to QS7YesYes
Run time< 30 min runs96-well: < 30 min; 384-well: < 35 min96-well: < 30 min; 384-well: < 35 min96-well: < 30 min; 384-well: < 35 min96-well: < 30 min; 384-well: < 35 min
VeriFlex™ block technologyYesYesYesYesNo
Gradient capacityVeriFlex™ 3 Zone (5°C zone-to-zone)VeriFlex™ 6 Zone (5°C zone-to-zone)VeriFlex™ 3 Zone (5°C zone-to-zone)VeriFlex™ 6 Zone (5°C zone-to-zone)N/A
Filter/colors4 coupled excitation/emission filters6 decoupled excitation/emission filters (0.1 or 0.2 mL)5 coupled excitation/emission filters6 decoupled excitation/emission filters (21 filter combos)6 coupled excitation/emission filters (21 filter combos)
Dye compatibility*FAM™/SYBR™ Green; VIC™/JOE™/HEX™/TET™; ABY™/NED™/TAMRA™/Cy®3; JUN™/ROX™/Texas Red™; custom dyesSame as QS3 plus Mustang Purple™, Cy®5, LIZ™, Alexa Fluor™ 647, Cy®5.5 (96-well only); custom dyesSame as QS5 (without Alexa Fluor™ 647 note); custom dyesSame as QS6 Pro; custom dyesSame as QS7 Pro; custom dyes
21 CFR Part 11 supportNoYesNoYesYes
Footprint (W × D × H)27 × 50 × 40 cm27 × 50 × 40 cm33.8 × 52.5 × 54.7 cm33.8 × 52.5 × 54.7 cm53 × 70 × 75 cm
Touchscreen8.4-inch capacitive8.4-inch capacitive12-inch capacitive12-inch capacitive10.4-inch capacitive
System configurationStand-alone, PC connected, or LAN/Wi-Fi via Connect PlatformStand-alone, PC connected, or LAN/Wi-Fi via Connect PlatformStand-alone, PC connected, or LAN/Wi-Fi via Connect PlatformStand-alone, PC connected, or LAN/Wi-Fi via Connect PlatformPC connected, LAN
Dynamic range10 logs10 logs10 logs10 logs10 logs (OpenArray™: 7 logs)
Analytical sensitivity1 copy1 copy1 copy1 copy1 copy
Discrimination / resolution1.5-fold concentrations1.5-fold concentrations1.5-fold concentrations1.5-fold concentrations1.5-fold (OpenArray™: 2-fold)