UserName
Applied Biosystems Gene Expression Tools
Applied Biosystems Gene Expression Tools
When choosing your real-time PCR master mixes, it’s important to make a selection that’s optimized for your application. Applied Biosystems™ products deliver the best-in-class performance, speed, and flexibility you need with straightforward protocols and integrated workflows.
  • Applied Biosystems™ PowerTrack™ SYBR™ Green Master Mixes are designed for quantitative real-time PCR using a set of two primers that flank the target region. These master mixes contain buffer, dNTPs, thermostable hot-start DNA polymerase, and, of course, SYBR green dye. Simply add your sample and PCR primer pair to get started.
  • Applied Biosystems™ TaqMan® Fast Advanced Master Mixes contain buffer, dNTPs, passive reference dye, thermostable hot-start DNA polymerase, and other components for reliable assay-based, real-time PCR. All you need to add is your sample and a TaqMan assay.
Find the Right Platform for Your Research
Applied Biosystems™ QuantStudio™ Real-Time PCR Systems
Request a Sample
Complete this form to request a sample of Applied Biosystems PowerTrack SYBR Green Master Mix or Applied Biosystems TaqMan Fast Advanced Master Mix.
Required Field *
Required Field *
Country
State/Province
By submitting your data, you agree to our Privacy Policy and will receive emails based on your preferences.
© Thermo Fisher Scientific, Inc. All Rights Reserved.
Thermo Fisher Scientific Logo
Select Detection Chemistry Products for Your Workflow
SYBR Green-Based Detection
SYBR Green-Based Detection
Applied Biosystems PowerTrack SYBR Green Master Mix
Specially crafted to work with SuperScript IV VILO Reverse Transcriptase for a faster and easier workflow, PowerTrack SYBR Green Master Mix also accommodates broad primer melting temperatures for consistency across qPCR runs.
TaqMan Probe-Based Detection
Three Easy Steps
Find Your Assay
TaqMan Real-Time Assays
TaqMan Real-Time Assays
Generate reliable data while saving time and money with Applied Biosystems™ TaqMan® real-time PCR assays. Choose from several formats, over 20 million assays, and more than 28 species.
Choose Your Control
TaqMan Real-Time PCR Controls
TaqMan Real-Time PCR Controls
Rapidly identify the optimal control for your assay with reliable, cost-effective Applied Biosystems TaqMan Endogenous Controls. Test as many or as few samples as needed with these controls, available in multiple formats.
Select Your Master Mix
TaqMan Fast Advanced Master Mix
TaqMan Fast Advanced Master Mix
Get accurate quantification in less time across an array of qPCR instrument platforms with Applied Biosystems TaqMan Fast Advanced Master Mix. It delivers higher sensitivity, greater specificity, and faster results.