Filtered Search Results
Fisherbrand™ Traceable™ Vaccine Refrigerator/Freezer Thermometer
Monitors temperature of vaccine in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor.
| Height (Metric) | 19 mm |
|---|---|
| Length (Metric) Cable | 25.4 cm |
| Length (Metric) | 70 mm |
| Length (English) | 2.75 in. |
| Min. Temperature (English) | -58°F |
| Material | ABS Plastic |
| Bottle Fill | Glycol |
| Max. Temperature (Metric) | 70°C |
| For Use With (Application) | Monitoring temperature in freezers, water baths, heating blocks, incubators and refrigerators |
| Battery Type | AA Alkaline (1) |
| Accuracy | ±1°C |
| For Use With (Equipment) | Refrigerators/Freezers |
| Product Type | Fridge/Freezer Digital Thermometer |
| Max. Temperature (English) | 158°F |
| Temperature Range (English) | -58°F to +158°F |
| Length (English) Cable | 10 in. |
| Weight (English) | 4 oz. |
| Certifications/Compliance | ISO 17025, A2LA, NIST |
| Temperature Range (Metric) | -50°C to +70°C |
| Min. Temperature (Metric) | -50°C |
| Calibrated | Yes |
| Display | Digital LCD |
| Resolution | 1°C |
| Height (English) | 0.75 in. |
Fisherbrand™ Traceable™ Excursion-Trac™ Datalogging Thermometer
Traceable™ Excursion-Trac™ Datalogging Thermometer with user-defined time intervals.
| Data Memory | 525,600 Readings |
|---|---|
| Software Type | TraceableLIVE |
| Sensor Types | Temperature-Buffered Bottle Sensor |
| Calibration | Calibrated |
| Alarm Types | LED Indicator,Audible,Flashing LCD Segment |
| Type | Datalogger |
| Display Type | LCD |
| Length (English) Cable | 10 ft. (Cable) |
| Temperature Range (English) | -58° to +158°F |
| Certifications/Compliance | ISO 17025, A2LA, NIST |
| Temperature Range (Metric) | -50.00°C to +70.00°C |
| Accuracy (Temperature) | ±0.25°C |
| Interface | USB |
| Temperature Resolution | 0.01° |
Fisherbrand™ Traceable Circle Laser Infrared Thermometer with Type K and Calibration; 12:1 Ratio
Measure and monitor two readings simultaneously
Fisherbrand™ Traceable™ Refrigerator/Freezer Alarm Thermometers
Provides accurate reading when freezer/refrigerator doors are open with temperature-buffered, sensor-sealed in bottled glycol solution
Fisherbrand™ Traceable™ Noncontact Infrared Thermometer
Traceable™ Noncontact Infrared Thermometer measures infrared surface temperatures with view fields of 6:1 to 8:1.
| Certifications/Compliance | ISO 17025, A2LA, NIST |
|---|---|
| For Use With (Application) | Petroleum, electronics, plant and field use. (Not meant for medical use.) |
| Measurement Time | 1 sec. |
| Display | Digital LCD |
Fisherbrand™ Traceable™ Digital Thermometer with Stainless-Steel Stem and 0.25 in. LCD Screen
Traceable™ Digital Thermometer used to measure temperature of gases, food, and semisolids.
| Product Type | Digital Thermometer |
|---|---|
| Min. Temperature (English) | -58°F |
| Material | Stainless Steel |
| Min. Temperature (Metric) | -50°C |
| Calibrated | Yes |
| Display | Digital LCD |
Fisherbrand™ Traceable™ Dishwasher Thermometer with Stainless-Steel Probe
Reports the maximum and minimum temperatures achieved during the dishwashing cycle. Traceable™ Dishwasher Thermometer with Stainless-Steel Probe ensures proper cleaning by providing proof of meeting the temperature regulation of achieving 180°F (82°C) during dishwashing.
Fisherbrand™ Traceable™ Total-Range Thermometer
Technologically advanced, pocket-size thermometer with water-resistant case
Fisherbrand™ Traceable™ Digital Thermometers with Short Sensors
Fisherbrand™ Traceable™ Digital Thermometers are ideal for monitoring liquids, air/gas or semisolids.
| Accuracy | ±1°C |
|---|---|
| Calibrated | Yes |
| Display | Digital LCD |
Fisherbrand™ Digital Thermometers with Stainless-Steel Probe on Cable
Easy-to-read digital thermometers are compact to hold in your hand or mount on wall in restricted areas
| Product Type | Traceable Digital Thermometer w/Probe |
|---|---|
| Certifications/Compliance | ISO 17025, A2LA, NIST |
| Calibrated | Yes |
| Display | Digital LCD |
Fisherbrand™ Traceable™ Dual-Channel Thermometer
Traceable™ Dual-Channel Thermometer digital display shows temperatures in °C or °F.
Fisherbrand™ Traceable™ Hi-Accuracy Refrigerator Thermometer
Traceable™ Hi-Accuracy Refrigerator Thermometer can monitor two different areas simultaneously. Has the ability to monitor temperatures in refrigerator and freezer simultaneously or two refrigerator locations.
| Width (English) | 2.75 in. |
|---|---|
| Length (Metric) Cable | 304.8 cm |
| Length (Metric) | 114 mm |
| Length (English) | 4.5 in. |
| Min. Temperature (English) | -58°F |
| For Use With (Application) | Monitoring temperature in refrigerator and freezer simultaneously or two refrigerator locations |
| Width (Metric) | 6.9 cm |
| Accuracy | ±0.3°C |
| For Use With (Equipment) | Refrigerators, Freezers |
| Max. Temperature (English) | 158°F |
| Length (English) Cable | 120 in. |
| Certifications/Compliance | ISO 17025, A2LA, NIST |
| Min. Temperature (Metric) | -50°C |
| Calibrated | Yes |
| Weight (Metric) | 140 g |
| Diameter (Metric) Probe | 4.57 mm |
| Material | ABS Plastic |
| Length (English) Probe | 0.7 in. |
| Max. Temperature (Metric) | 70°C |
| Length (Metric) Probe | 1.9 cm |
| Product Type | Digital Thermometer |
| Diameter (English) Probe | 0.18 in. |
| Temperature Range (English) | -58°F to +158°F |
| Weight (English) | 5 oz. |
| Temperature Range (Metric) | -50°C to +70°C |
| Display | LED |
| Resolution | 0.01°C |
Fisherbrand™ TraceableLIVE™ Wi-Fi Datalogging Refrigerator/Freezer Thermometer with Remote Notification
TraceableLIVE™ Wi-Fi Datalogging Refrigerator/Freezer Thermometer now with FREE TraceableLIVE™ subscription plan (for one user). A simple, efficient, and reliable way to stay connected to your critical environments with smartphone, tablet or personal computer.
| Alarm Types | Mobile Push Notification,E-mail,SMS |
|---|---|
| Type | Datalogger |
| Display Type | Digital |
| Software Type | Web, Android, Apple IOS |
| Temperature Range (English) | -58°F to +140°F |
| Calibration | Multiple-point Temperature Calibration |
| Temperature Range (Metric) | -50°C to +60°C |
| Accuracy (Temperature) | ±0.25°C (-40°C to +60°C), ±0.50°C otherwise |
| Interface | Cloud-Based Data Interface |
| Temperature Resolution | 0.01° |
Fisherbrand™ TraceableLIVE™ Ultra-Low Temperature WiFi Datalogging Thermometer with Remote Notification
With FREE TraceableLIVE™ 30-day trial subscription (for one user). A simple, efficient, and reliable way to stay connected to your critical environments with smartphone, tablet or personal computer.
| Alarm Types | Mobile Push Notification,E-mail,SMS |
|---|---|
| Type | Temperature Datalogger |
| Display Type | Digital |
| Software Type | TraceableLIVE |
| Certifications/Compliance | Traceable Certificate which assures accuracy from an ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA |
| Calibration | Multiple-point Temperature Calibration |
| Temperature Range (Metric) | -90.00°C to +105.00°C |
| For Use With (Application) | Remotely monitor temperature |
| Accuracy (Temperature) | ±0.1°C |
| Interface | Cloud-Based Data Interface |
| Temperature Resolution | 0.01° |
Fisherbrand™ TraceableLIVE™ WiFi Datalogging Hygrometer/Thermometer with Remote Notification
TraceableLIVE™ WiFi Datalogging Hygrometer/Thermometer now with FREE TraceableLIVE subscription plan (for one user). Stay connected to your lab with your smartphone, tablet, or personal computer. View current temperatures and humidities, control alarm parameters, view datalogging history, and generate reports by using TraceableLIVE™ app.
| Type | Hygrometer Datalogger |
|---|---|
| Resolution (Relative Humidity) | 0.01% |
| Accuracy (Relative Humidity) | ±3% RH [20 to 80] %RH, ±5% RH otherwise |
| Display Type | Digital |
| Humidity Range | 0 to 100% |
| Accuracy (Temperature) | ±0.4°C |
| Temperature Resolution | 0.01° |