missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ TraceableLIVE™ Ultra-Low Temperature WiFi Datalogging Thermometer with Remote Notification

Catalog No. p-6824015
Change view
Click to view available options
Includes:
1 Stainless-steel 316 Probe, Quick Start Guide, batteries, magnets, Velcro, wall mount, and Traceable™ Certificate
2 Stainless-steel 316 Probes, Quick Start Guide, batteries, magnets, Velcro, wall mount, and Traceable™ Certificate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
15079627 2 Stainless-steel 316 Probes, Quick Start Guide, batteries, magnets, Velcro, wall mount, and Traceable™ Certificate
15079626 1 Stainless-steel 316 Probe, Quick Start Guide, batteries, magnets, Velcro, wall mount, and Traceable™ Certificate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15079627 Supplier Fisherbrand™ Supplier No. 6101144

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Requires

TracebleLIVE requires subscription

With FREE TraceableLIVE™ 30-day trial subscription (for one user). A simple, efficient, and reliable way to stay connected to your critical environments with smartphone, tablet or personal computer.

A simple, efficient, and reliable way to stay connected to your critical environments with smartphone, tablet or personal computer.

Use the TraceableLIVE™ app†, or visit Traceablelive.com† on any browser to view your current temperatures, control your alarm parameters, view your datalogging history, view and run reports or give other team members access to the data.

  • Remotely monitor temperature and more by connecting to existing WiFi
  • Alerts: temperature alarm, connectivity interruption, low battery
  • Cloud-based data interface that requires no local software; web, Android, Apple IOS data visibility
  • Continuous data transmission and monitoring using SSL encrypted SECURE connection
  • Receive Mobile Push/E-Mail/Text notifications
  • Unlimited scalability - use TraceableLIVE™ in individual labs or an entire network of facilities
  • Multiple point temperature calibration traceable to NIST

†Requires TraceableLIVE subscription

TraceableLIVE Subscription Cost through traceablelive.com:

  • After 30-day trial, user will be required to purchase the software for continued use
  • Monthly Payment Plan: $8.99 per device for 1-9 devices or $7.20 per device for 10+ devices
  • Yearly Payment Plan: $100 per device for 1-9 devices or $80 per device for 10+ devices
  • Demo TraceableLive software at traceablelive.com

Note: To order Standard or Premium Subscription Coupon Codes, contact Fisher Customer Service.

Specifications

Type Temperature Datalogger
Display Type Digital
Calibration Multiple-point Temperature Calibration
Alarm Types Mobile Push Notification, E-mail, SMS
Software Type TraceableLIVE
Includes 2 Stainless-steel 316 Probes, Quick Start Guide, batteries, magnets, Velcro, wall mount, and Traceable™ Certificate
Certifications/Compliance Traceable Certificate which assures accuracy from an ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA
Interface Cloud-Based Data Interface
For Use With (Application) Remotely monitor temperature
Accuracy (Temperature) ±0.1°C
Temperature Resolution 0.01°
Temperature Range (Metric) -90.00°C to +105.00°C
Show More Show Less