missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Refrigerator/Freezer Alarm Thermometers

Catalog No. 0666423
Change view
Click to view available options
Temperature Range (Metric):
-50°C to +70°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
0666423 -50°C to +70°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0666423
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Velcro™ and magnetic mounting strips, one AAA battery, and ABS plastic case

Provides accurate reading when freezer/refrigerator doors are open with temperature-buffered, sensor-sealed in bottled glycol solution

Provides accurate reading when freezer/refrigerator doors are open with temperature-buffered, sensor-sealed in bottled glycol solution. Fisher Scientific™ Traceable™ Refrigerator/Freezer Alarm Thermometer shows current, high and low temperatures and alerts user with visual and audible signals if temperature diverts from set points.

  • Monitors temperature in freezers, refrigerators, incubators, water baths and heating blocks
  • Triple display simultaneously shows current, high and low temperatures
  • Alarm (Cat. No. 06-664-11 only) set in 1° increments alerts user with visual and audible signals if temperature goes outside setpoints
  • 10 ft. long (3m) micro cable permits doors to close on it
  • Dimensions: 4.25L x 2.75W x 0.75 in.H (11 x 7 x 2cm)

Certifications

Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.

Specifications

Certifications/Compliance ISO 17025, A2LA, NIST
Dimensions (L x W x H) 2.5 x 3.25 x 0.63 in. (63.5 x 82.5 x 16 mm)
For Use With (Application) Monitoring temperature in freezers, refrigerators, incubators, heating blocks
Height (English) 0.63 in.
Height (Metric) 16 mm
Includes Velcro™ and magnetic mounting strip, one AAA battery, ABS Plastic case
Length (English) Overall 2.5 in.
Length (Metric) 63.5 mm
Length (Metric) Overall 63.5 mm
Material Plastic
Resolution 0.1°
Weight (English) 4 oz.
Weight (Metric) 0.11 kg
Width (English) 3.25 in.
Product Type Refrigerator/Freezer Thermometer
Description Bottle Probe
Max. Temperature (English) 158°F
Max. Temperature (Metric) 70°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +158°F
Temperature Range (Metric) -50°C to +70°C
Calibrated Yes
For Use With (Equipment) Monitoring Temperature in Freezer, Refrigerator, Incubator, Heating Block
Accuracy ±0.5°C
Display Digital LCD
Width (Metric) 82.5 mm
Show More Show Less