missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Excursion-Trac™ Datalogging Thermometer

Catalog No. p-4700702
Change view
Click to view available options
Includes:
1 Bottle Probe
1 Bullet Probe (5/16 in. dia. x 3/4 in. long)
1 Glass Vaccine Bottle Probe
1 Plastic Bottle Probe
1 Stainless-Steel Probe
2 Bottle Probes
2 Glass Vaccine Bottle Probes
2 Plastic Bottle Probes
2 Probes (Bottle Probe and 5/16 in. dia. x 3/4 in. long)
2 Probes, 5/16 in. dia. x 3/4 in. long
2 Stainless-Steel Probes
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
15081123 1 Bottle Probe
15081124 2 Bottle Probes
15081125 1 Bullet Probe (5/16 in. dia. x 3/4 in. long)
15081126 2 Probes, 5/16 in. dia. x 3/4 in. long
15081127 2 Probes (Bottle Probe and 5/16 in. dia. x 3/4 in. long)
15081128 1 Stainless-Steel Probe
15081129 2 Stainless-Steel Probes
15079656 1 Glass Vaccine Bottle Probe
15079657 2 Glass Vaccine Bottle Probes
15079660 1 Plastic Bottle Probe
15079661 2 Plastic Bottle Probes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 15081123
Only null left

Includes

Stand, Velcro™, magnetic strips, wall mount, AC power adapter, batteries, and an ISO 17025 Traceable™ Certificate

Traceable™ Excursion-Trac™ Datalogging Thermometer with user-defined time intervals.

High-accuracy thermometer has a temperature range of -50 to 70°C and resolution of 0.01°C and maintains an accuracy of ±0.25°C across the entire temperature range.

  • Stores up to ten unique alarm events
  • Meets current CDC requirements for vaccine storage and monitoring
  • Rolling memory structure, maintains most recent 525,600 temperature observations
  • Memory can be cleared on unit after downloading data to USB stick
  • Hassle-free retrieval of data; unit can remain in use while downloading and analyzing data
  • Status indicators: Low battery, memory full, USB data transfer and active alarm state
  • Traceable to NIST for accuracy
  • High-impact, chemical-resistant ABS plastic case measures 7 x 10.8 x 1.9 cm (2.75 x 4.25 x 0.75 in.)
  • Datalogging Capacity: 525,600 temperature observations
  • Weight: 5 oz.

Hi/Lo Alarms and Time/Date Stamps

  • Alarm tracking feature stores in memory each alarm event detailing when the alarm state occurred, as well as when the unit returned back to within range
  • Alarm state indicators include visual LEDs, audio alerts, and flashing LCD segments
  • Data for up to ten different alarm events can be retrieved with the most current event viewable on the display

USB Data Transferring

  • New feature allows recorded data (CSV file) to be transferred from thermometer to PC or Mac™ using a USB flash drive (not included)
  • No software is needed for computer interface
  • Each thermometer has a unique ID allowing multiple units to be used in the same location

Data-Logging Features

  • Monitor readings overnight, on weekends, or for any time period with rolling data log
  • User-defined data-logging interval of one observation/minute to one observation/24 hours
  • Memory capacity maintains a year of recorded data when using one-minute intervals and longer when using larger intervals

Bottle Probe Features (Cat. Nos. 15-081-125, -126, -128, 129 do not come with bottle probes)

  • Bottle probes are sealed in a miniature bottle (1 x 2.5 in.) filled with nontoxic glycol
  • Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration)
  • Eliminates concerns about incidental contact with food or drinking water
  • Temperature-buffered bottle sensor eliminates rapid changes when refrigerator door is opened
  • Ideal for use in monitoring storage conditions for vaccines or other important samples

Note: Units are designed to be used with either the provided AC adaptor or with batteries only. Batteries are designed to last for up to six months. If using battery-power only, the AC power adaptor must be plugged in to download any data.

Order Info

Replacement battery: Cat. No. 1111

Certifications

Supplied with an ISO 17025 Traceable™ Certificate

Specifications

Type Datalogger
Display Type LCD
Data Memory 525,600 Readings
Calibration Calibrated
Alarm Types LED Indicator, Audible, Flashing LCD Segment
Software Type TraceableLIVE
Description 1 Bottle Probe
Includes 1 Bottle Probe
Certifications/Compliance ISO 17025, A2LA, NIST
Interface USB
Sensor Types Temperature-Buffered Bottle Sensor
For Use With (Application) Monitoring temperatures in refrigerators, freezers, water baths, heating blocks, and incubators.
Accuracy (Temperature) ±0.25°C
Resolution 0.01°
Response Time User-Defined Timing Interval: Yes
Temperature Resolution 0.01°
Temperature Range (English) -58° to +158°F
Temperature Range (Metric) -50.00°C to +70.00°C
Length (English) Cable 10 ft. (Cable)
Show More Show Less