CAS RN 95-47-6
o-Xylene (Certified), Fisher Chemical
CAS: 95-47-6 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008519 InChI Key: CTQNGGLPUBDAKN-UHFFFAOYSA-N Synonym: o-xylene,1,2-dimethylbenzene,ortho-xylene,o-xylol,o-methyltoluene,o-dimethylbenzene,2-xylene,3,4-xylene,benzene, 1,2-dimethyl,o-xylenes PubChem CID: 7237 ChEBI: CHEBI:28063 IUPAC Name: 1,2-xylene SMILES: CC1=CC=CC=C1C
o-Xylene, HPLC Grade, 96% min
CAS: 95-47-6 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008519 InChI Key: CTQNGGLPUBDAKN-UHFFFAOYSA-N Synonym: o-xylene,1,2-dimethylbenzene,ortho-xylene,o-xylol,o-methyltoluene,o-dimethylbenzene,2-xylene,3,4-xylene,benzene, 1,2-dimethyl,o-xylenes PubChem CID: 7237 ChEBI: CHEBI:28063 IUPAC Name: 1,2-xylene SMILES: CC1=CC=CC=C1C
o-Xylene, 99%
CAS: 95-47-6 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008519 InChI Key: CTQNGGLPUBDAKN-UHFFFAOYSA-N Synonym: o-xylene,1,2-dimethylbenzene,ortho-xylene,o-xylol,o-methyltoluene,o-dimethylbenzene,2-xylene,3,4-xylene,benzene, 1,2-dimethyl,o-xylenes PubChem CID: 7237 ChEBI: CHEBI:28063 IUPAC Name: 1,2-xylene SMILES: CC1=CC=CC=C1C
o-Xylene, Spectrophotometric Grade, 96% min
CAS: 95-47-6 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008519 InChI Key: CTQNGGLPUBDAKN-UHFFFAOYSA-N Synonym: o-xylene,1,2-dimethylbenzene,ortho-xylene,o-xylol,o-methyltoluene,o-dimethylbenzene,2-xylene,3,4-xylene,benzene, 1,2-dimethyl,o-xylenes PubChem CID: 7237 ChEBI: CHEBI:28063 IUPAC Name: 1,2-xylene SMILES: CC1=CC=CC=C1C
o-Xylene, 99%, Extra Dry, AcroSeal™
CAS: 95-47-6 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008519 InChI Key: CTQNGGLPUBDAKN-UHFFFAOYSA-N Synonym: o-xylene,1,2-dimethylbenzene,ortho-xylene,o-xylol,o-methyltoluene,o-dimethylbenzene,2-xylene,3,4-xylene,benzene, 1,2-dimethyl,o-xylenes PubChem CID: 7237 ChEBI: CHEBI:28063 IUPAC Name: 1,2-xylene SMILES: CC1=CC=CC=C1C
o-Xylene, Reagent Grade, ≥98.0%, Solstice
CAS: 95-47-6 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008519 InChI Key: CTQNGGLPUBDAKN-UHFFFAOYSA-N Synonym: o-xylene,1,2-dimethylbenzene,ortho-xylene,o-xylol,o-methyltoluene,o-dimethylbenzene,2-xylene,3,4-xylene,benzene, 1,2-dimethyl,o-xylenes PubChem CID: 7237 ChEBI: CHEBI:28063 IUPAC Name: 1,2-xylene SMILES: CC1=CC=CC=C1C
o-Xylene, 99%, pure
CAS: 95-47-6 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008519 InChI Key: CTQNGGLPUBDAKN-UHFFFAOYSA-N Synonym: o-xylene,1,2-dimethylbenzene,ortho-xylene,o-xylol,o-methyltoluene,o-dimethylbenzene,2-xylene,3,4-xylene,benzene, 1,2-dimethyl,o-xylenes PubChem CID: 7237 ChEBI: CHEBI:28063 IUPAC Name: 1,2-xylene SMILES: CC1=CC=CC=C1C
O-Xylene, Certified Reference Material, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.
o-Xylene 98.0+%, TCI America™
CAS: 95-47-6 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008519 InChI Key: CTQNGGLPUBDAKN-UHFFFAOYSA-N Synonym: o-xylene,1,2-dimethylbenzene,ortho-xylene,o-xylol,o-methyltoluene,o-dimethylbenzene,2-xylene,3,4-xylene,benzene, 1,2-dimethyl,o-xylenes PubChem CID: 7237 ChEBI: CHEBI:28063 IUPAC Name: 1,2-xylene SMILES: CC1=CC=CC=C1C
o-Xylene, SPEX CertiPrep™
CAS: 95-47-6 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008519 InChI Key: CTQNGGLPUBDAKN-UHFFFAOYSA-N PubChem CID: 7237 ChEBI: CHEBI:28063 IUPAC Name: 1,2-xylene SMILES: CC1=CC=CC=C1C
o-Xylene, 98.0%, MilliporeSigma™
CAS: 95-47-6 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 InChI Key: CTQNGGLPUBDAKN-UHFFFAOYSA-N IUPAC Name: 1,2-xylene SMILES: CC1=CC=CC=C1C
o-Xylene, CHROMASOLV™ Plus, for HPLC, 98%, Solstice
CAS: 95-47-6 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008519 InChI Key: CTQNGGLPUBDAKN-UHFFFAOYSA-N Synonym: o-xylene,1,2-dimethylbenzene,ortho-xylene,o-xylol,o-methyltoluene,o-dimethylbenzene,2-xylene,3,4-xylene,benzene, 1,2-dimethyl,o-xylenes PubChem CID: 7237 ChEBI: CHEBI:28063 IUPAC Name: 1,2-xylene SMILES: CC1=CC=CC=C1C
HYDRANAL™ - Xylene, Solvent for KF titration, Solstice
Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008519 InChI Key: CTQNGGLPUBDAKN-UHFFFAOYSA-N Synonym: 1,2-Dimethylbenzene PubChem CID: 7237 ChEBI: CHEBI:28063 IUPAC Name: 1,2-xylene SMILES: CC1=CC=CC=C1C
- 1
- 2