missing translation for 'onlineSavingsMsg'
Learn More

o-Xylene, EMD-Millipore

Catalog No. M1097980005
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1097980005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1097980005 Supplier Technidata Supplier No. 1.09798.0005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hydrocarbons C8

Chemical Identifiers

CAS 95-47-6
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008519
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
Synonym 1, 2-Dimethylbenzene
PubChem CID 7237
ChEBI CHEBI:28063
IUPAC Name 1,2-xylene
SMILES CC1=CC=CC=C1C

Specifications

Packaging Glass ampoule
Explosion Limit 1.0 to 7.6 %(V)
Ignition Point 465°C
Vapor Pressure 7 hPa (20°C)
Density Density: 0.88g/cm3 (20°)
Assay Percent Range ≥99.0%
Linear Formula C6H4(CH3)2
Quantity 5 mL
Formula Weight 106.17
Chemical Name or Material o-Xylene
Recommended Storage Store without restrictions