missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Xylene, Certified Reference Material, MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | o-Xylene |
| Melting Point | -26°C to -23°C (literature) |
| CAS | 95-47-6 |
| Density | 0.879 g/mL (at 20°C (literature)) |
| Quantity | 20 mL |
| Flash Point | 31°C (Not Applicable) |
| Refractive Index | n20/D 1.505 (literature) |
| Molecular Formula | C8H10 |
| Linear Formula | C6H4(CH3)2 |
| Vapor Pressure | 16 mmHg (37.7°C) |
| Show More |