missing translation for 'onlineSavingsMsg'
Learn More

o-Xylene, Reagent Grade, ≥98.0%, Solstice

Catalog No. 6001152
Change view
Click to view available options
Quantity:
18 L
Packaging:
Stainless Steel Can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001152 18 L Stainless Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001152 Supplier Solstice Supplier No. X104018L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent Grade,≥98.0%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 95-47-6
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008519
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
Synonym o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
PubChem CID 7237
ChEBI CHEBI:28063
IUPAC Name 1,2-xylene
SMILES CC1=CC=CC=C1C

Specifications

Density 0.879g/mL at 20°C
Quantity 18 L
Explosion Limit 0.07
Flash Point 31°C (87.8°F)
Packaging Stainless Steel Can
Vapor Pressure 16 mmHg (37.7°C), 0.1 atm (21.1°C), 7 mmHg (20°C)
Linear Formula C6H4(CH3)2
UN Number UN1307
Beilstein 1815558
Formula Weight 106.17g/mol
Percent Purity ≥98%
Grade Not Available
Chemical Name or Material o-Xylene
Show More Show Less
Hazard Statement P210-P261-P273-P301 + P310-P302 + P352 + P312-P331
RTECSNumber ZE2450000