missing translation for 'onlineSavingsMsg'
Learn More

o-Xylene, Analytical Standard, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019790
Change view
Click to view available options
Quantity:
5 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019790 5 mL
111018214 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019790 Supplier Merck Emd Millipore Supplier No. 956605ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 95-47-6
Vapor Pressure 16 mmHg (37.7°C)
Vapor Density 3.7 (vs. air)
Density 0.879 g/mL (at 20°C)
Physical Form Neat
Linear Formula C6H4(CH3)2
MDL Number MFCD00008519
Melting Point -26°C to +23°C
Boiling Point 143°C to 145°C
Synonym 1,2-Dimethylbenzene
Quantity 5 mL
Formula Weight 106.17
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less