missing translation for 'onlineSavingsMsg'
Learn More

o-Xylene, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019591
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019591 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019591 Supplier Merck Emd Millipore Supplier No. PHR13121G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

o-Xylene is a nonoxygenated, homocyclic aromatic compound that occurs as a constituent of petroleum. It belongs to the class of volatile organic compounds (VOCs) having vapor pressure high enough to vaporize under normal conditions and enter into the atmosphere. It is found to be relatively water-soluble and suspected to have carcinogenic properties.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 95-47-6
Vapor Pressure 16 mmHg (37.7°C)
Vapor Density 3.7 (vs. air)
Density 0.879 g/mL (at 20°C)
Physical Form Neat
Linear Formula C6H4(CH3)2
MDL Number MFCD00008519
Melting Point -26°C to +23°C
Boiling Point 143°C to 145°C
Synonym 1,2-Dimethylbenzene
Quantity 1 g
Formula Weight 106.17
Grade Certified Reference Material
Show More Show Less