CAS RN 87-89-8
myo-Inositol, 99%
CAS: 87-89-8 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 MDL Number: MFCD00077932 InChI Key: CDAISMWEOUEBRE-UHFFFAOYSA-N Synonym: scyllo-inositol,myo-inositol,inositol,muco-inositol,epi-inositol,i-inositol,meso-inositol,allo-inositol,1d-chiro-inositol,myoinositol PubChem CID: 892 ChEBI: CHEBI:24848 IUPAC Name: cyclohexane-1,2,3,4,5,6-hexol SMILES: C1(C(C(C(C(C1O)O)O)O)O)O
Thermo Scientific Chemicals myo-Inositol, Cell Culture Grade
CAS: 87-89-8 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 MDL Number: MFCD00077932 InChI Key: CDAISMWEOUEBRE-UHFFFAOYSA-N Synonym: scyllo-inositol,myo-inositol,inositol,muco-inositol,epi-inositol,i-inositol,meso-inositol,allo-inositol,1d-chiro-inositol,myoinositol PubChem CID: 892 ChEBI: CHEBI:24848 IUPAC Name: cyclohexane-1,2,3,4,5,6-hexol SMILES: C1(C(C(C(C(C1O)O)O)O)O)O
Thermo Scientific Chemicals Inositol, 98+%
CAS: 87-89-8 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.16 MDL Number: MFCD00077932 InChI Key: CDAISMWEOUEBRE-UHFFFAOYSA-N Synonym: scyllo-inositol,myo-inositol,inositol,muco-inositol,epi-inositol,i-inositol,meso-inositol,allo-inositol,1d-chiro-inositol,myoinositol PubChem CID: 892 ChEBI: CHEBI:24848 IUPAC Name: cyclohexane-1,2,3,4,5,6-hexol SMILES: C1(C(C(C(C(C1O)O)O)O)O)O
Thermo Scientific Chemicals myo-Inositol, 98+%
CAS: 87-89-8 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 MDL Number: MFCD00077932 InChI Key: CDAISMWEOUEBRE-UHFFFAOYSA-N Synonym: scyllo-inositol,myo-inositol,inositol,muco-inositol,epi-inositol,i-inositol,meso-inositol,allo-inositol,1d-chiro-inositol,myoinositol PubChem CID: 892 ChEBI: CHEBI:24848 IUPAC Name: cyclohexane-1,2,3,4,5,6-hexol SMILES: C1(C(C(C(C(C1O)O)O)O)O)O
D-Myo-inositol, 99.26%, For HPLC analysis, MP Biomedicals™
CAS: 87-89-8 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 InChI Key: CDAISMWEOUEBRE-UHFFFAOYSA-N Synonym: scyllo-inositol,myo-inositol,inositol,muco-inositol,epi-inositol,i-inositol,meso-inositol,allo-inositol,1d-chiro-inositol,myoinositol PubChem CID: 892 ChEBI: CHEBI:24848 IUPAC Name: cyclohexane-1,2,3,4,5,6-hexol SMILES: C1(C(C(C(C(C1O)O)O)O)O)O
myo-Inositol 99.0+%, TCI America™
CAS: 87-89-8 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 MDL Number: MFCD00077932 InChI Key: CDAISMWEOUEBRE-UHFFFAOYSA-N Synonym: scyllo-inositol,myo-inositol,inositol,muco-inositol,epi-inositol,i-inositol,meso-inositol,allo-inositol,1d-chiro-inositol,myoinositol PubChem CID: 892 ChEBI: CHEBI:24848 IUPAC Name: cyclohexane-1,2,3,4,5,6-hexol SMILES: C1(C(C(C(C(C1O)O)O)O)O)O
D-myo-Inositol, 99.2%, Cell culture reagent, For HPLC analysis, MP Biomedicals™
CAS: 87-89-8 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.156 MDL Number: MFCD00077932 InChI Key: CDAISMWEOUEBRE-UHFFFAOYSA-N Synonym: scyllo-inositol,myo-inositol,inositol,muco-inositol,epi-inositol,i-inositol,meso-inositol,allo-inositol,1d-chiro-inositol,myoinositol PubChem CID: 892 ChEBI: CHEBI:24848 IUPAC Name: cyclohexane-1,2,3,4,5,6-hexol SMILES: C1(C(C(C(C(C1O)O)O)O)O)O
Inositol, 97%, Spectrum™ Chemical
CAS: 87-89-8 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.16 InChI Key: CDAISMWEOUEBRE-UHFFFAOYSA-N IUPAC Name: cyclohexane-1,2,3,4,5,6-hexol SMILES: OC1C(O)C(O)C(O)C(O)C1O
Inositol, FCC, 97%, Spectrum™ Chemical
CAS: 87-89-8 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.16 InChI Key: CDAISMWEOUEBRE-UHFFFAOYSA-N IUPAC Name: cyclohexane-1,2,3,4,5,6-hexol SMILES: OC1C(O)C(O)C(O)C(O)C1O
Inositol, NF, 97-102%, Spectrum™ Chemical
CAS: 87-89-8 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.16 InChI Key: CDAISMWEOUEBRE-UHFFFAOYSA-N IUPAC Name: cyclohexane-1,2,3,4,5,6-hexol SMILES: OC1C(O)C(O)C(O)C(O)C1O