missing translation for 'onlineSavingsMsg'
Learn More

Inositol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015736
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015736 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015736 Supplier Merck Emd Millipore Supplier No. PHR13511G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inositol is a structural isomer of glucose and a sugar alcohol. It is known to be a remedy for mild depression and neurasthenia.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 87-89-8
Vapor Density 6.2 (vs. air)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C6H12O6
MDL Number MFCD00077932
Melting Point 222°C to 227°C
Synonym myo-Inositol; 1,2,3,4,5,6-Hexahydroxycyclohexane; i-Inositol; meso-Inositol
Type Certified Reference Material
Quantity 1 g
Formula Weight 180.16
Grade Certified Reference Material
Show More Show Less