missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals myo-Inositol, 98+%

Catalog No. AAA1358636
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1358636 500 g
AAA1358622 100 g
AAA135860E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1358636 Supplier Thermo Scientific Chemicals Supplier No. A1358636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 87-89-8 | C6H12O6 | 180.156 g/mol

Myo-Inositol is a growth factor for animals and microorganisms. It is the most abundant form of polyols that serves as a structural element of secondary messengers in eukaryotic cells. It is used as an adulterant in many illegal drugs like cocaine and methamphetamine. Further, it is used as a stand-in for cocaine on television and film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Myo-Inositol is a growth factor for animals and microorganisms. It is the most abundant form of polyols that serves as a structural element of secondary messengers in eukaryotic cells. It is used as an adulterant in many illegal drugs like cocaine and methamphetamine. Further, it is used as a stand-in for cocaine on television and film.

Solubility
Soluble in water. Slightly soluble in ethanol, dimethyl sulfoxide and alcohol. Insoluble in ether.    

Notes
Incompatible with strong oxidizing agents.    

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00077932
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
Synonym scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
PubChem CID 892
ChEBI CHEBI:24848
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES C1(C(C(C(C(C1O)O)O)O)O)O

Specifications

Melting Point 224°C to 227°C
Density 1.75
Odor Odorless
Assay Percent Range 98%
Quantity 500 g
Beilstein 1907329
Merck Index 14,4978
Solubility Information Soluble in water. Slightly soluble in ethanol,dimethyl sulfoxide and alcohol. Insoluble in ether.
Formula Weight 180.16
Percent Purity ≥98%
Chemical Name or Material myo-Inositol
Show More Show Less
EINECSNumber 201-781-2
RTECSNumber NM7520800
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only