missing translation for 'onlineSavingsMsg'
Learn More

D-myo-Inositol, 99.2%, Cell culture reagent, For HPLC analysis, MP Biomedicals™

Catalog No. ICN19468850
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19468850 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19468850 Supplier MP Biomedicals Inc Supplier No. 0219468850

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00077932
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
Synonym scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
PubChem CID 892
ChEBI CHEBI:24848
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES C1(C(C(C(C(C1O)O)O)O)O)O

Specifications

Color White
Density 6.2g/mL
Quantity 50 g
Formula Weight 180.2
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material D-myo-inositol, Molecular biology and Cell biology
EINECSNumber 201-781-2
Recommended Storage Store at room temperature.