missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Inositol, 98+%

Catalog No. AC122265000
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122265000 500 g Plastic bottle
AC122261000 100 g Plastic bottle
AC122260010 1 kg Plastic bottle
AC122260051 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC122265000 Supplier Thermo Scientific Chemicals Supplier No. 122265000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 87-89-8 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00077932
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
Synonym scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
PubChem CID 892
ChEBI CHEBI:24848
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES C1(C(C(C(C(C1O)O)O)O)O)O

Specifications

Melting Point 220°C to 228°C
Color White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98% min. (ELSD) (HPLC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 06, II, 1157
Merck Index 15, 5021
Solubility Information (5% in water) Clear colorless
Formula Weight 180.16
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material Inositol, 98+%
Show More Show Less
EINECSNumber 201-781-2
RTECSNumber NM7520800
TSCA TSCA

RUO – Research Use Only