CAS RN 584-08-7
Potassium Carbonate Anhydrous (Granular Powder/Certified ACS), Fisher Chemical™
CAS: 584-08-7 Molecular Formula: CK2O3 Molecular Weight (g/mol): 138.21 MDL Number: MFCD00011382 InChI Key: BWHMMNNQKKPAPP-UHFFFAOYSA-L Synonym: potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate PubChem CID: 11430 IUPAC Name: dipotassium carbonate SMILES: [K+].[K+].[O-]C([O-])=O
Potassium carbonate, 98%, extra pure, anhydrous
CAS: 584-08-7 Molecular Formula: CK2O3 Molecular Weight (g/mol): 138.21 MDL Number: MFCD00011382 InChI Key: BWHMMNNQKKPAPP-UHFFFAOYSA-L Synonym: potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate PubChem CID: 11430 IUPAC Name: dipotassium;carbonate SMILES: [K+].[K+].[O-]C([O-])=O
Potassium carbonate, 99+%, ACS reagent, anhydrous
CAS: 584-08-7 Molecular Formula: CK2O3 Molecular Weight (g/mol): 138.21 MDL Number: MFCD00011382 InChI Key: BWHMMNNQKKPAPP-UHFFFAOYSA-L Synonym: potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate PubChem CID: 11430 SMILES: [K+].[K+].[O-]C([O-])=O
Potassium carbonate, anhydrous, 99%
CAS: 584-08-7 Molecular Formula: CK2O3 Molecular Weight (g/mol): 138.21 MDL Number: MFCD00011382 InChI Key: BWHMMNNQKKPAPP-UHFFFAOYSA-L Synonym: potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate PubChem CID: 11430 SMILES: [K+].[K+].[O-]C([O-])=O
Potassium Carbonate, 15% (w/v), Ricca Chemical
CAS: 584-08-7 Molecular Formula: CK2O3 Molecular Weight (g/mol): 138.21 MDL Number: MFCD00011382 InChI Key: BWHMMNNQKKPAPP-UHFFFAOYSA-L Synonym: potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate PubChem CID: 11430 IUPAC Name: dipotassium carbonate SMILES: [K+].[K+].[O-]C([O-])=O
Potassium carbonate, Puratronic™, 99.997% (metals basis)
CAS: 584-08-7 Molecular Formula: CK2O3 Molecular Weight (g/mol): 138.21 MDL Number: MFCD00011382 InChI Key: BWHMMNNQKKPAPP-UHFFFAOYSA-L Synonym: potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate PubChem CID: 11430 SMILES: [K+].[K+].[O-]C([O-])=O
Potassium carbonate, purum p.a., Solstice
CAS: 584-08-7 Molecular Formula: CK2O3 Molecular Weight (g/mol): 138.21 MDL Number: MFCD00011382 InChI Key: BWHMMNNQKKPAPP-UHFFFAOYSA-L Synonym: potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate PubChem CID: 11430 IUPAC Name: dipotassium;carbonate SMILES: [K+].[K+].[O-]C([O-])=O
Potassium carbonate, Reagent grade, Solstice
CAS: 584-08-7 Molecular Formula: CK2O3 Molecular Weight (g/mol): 138.21 MDL Number: MFCD00011382 InChI Key: BWHMMNNQKKPAPP-UHFFFAOYSA-L Synonym: potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate PubChem CID: 11430 SMILES: [K+].[K+].[O-]C([O-])=O
Potassium carbonate, 99+%, for analysis, anhydrous
CAS: 584-08-7 Molecular Formula: CK2O3 Molecular Weight (g/mol): 138.21 MDL Number: MFCD00011382 InChI Key: BWHMMNNQKKPAPP-UHFFFAOYSA-L Synonym: potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate PubChem CID: 11430 SMILES: [K+].[K+].[O-]C([O-])=O
Potassium carbonate, ACS, 99.0% min
CAS: 584-08-7 Molecular Formula: CK2O3 Molecular Weight (g/mol): 138.21 MDL Number: MFCD00011382 InChI Key: BWHMMNNQKKPAPP-UHFFFAOYSA-L Synonym: potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate PubChem CID: 11430 SMILES: [K+].[K+].[O-]C([O-])=O
Metals in Sewage Sludge - PT, MilliporeSigma™ Supelco™
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
Metals in Soil - PT, MilliporeSigma™ Supelco™
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
Potassium Carbonate, puriss p.a, ACS, ≥99.0% (T), Solstice
CAS: 584-08-7 Molecular Formula: CK2O3 Molecular Weight (g/mol): 138.21 MDL Number: MFCD00011382 InChI Key: BWHMMNNQKKPAPP-UHFFFAOYSA-L Synonym: potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate PubChem CID: 11430 IUPAC Name: dipotassium;carbonate SMILES: [K+].[K+].[O-]C([O-])=O
Potassium Carbonate, Saturated at 20°C, Reagents
CAS: 584-08-7 Molecular Formula: CK2O3 Molecular Weight (g/mol): 138.21 MDL Number: MFCD00011382 InChI Key: BWHMMNNQKKPAPP-UHFFFAOYSA-L Synonym: Dipotassium Carbonate, Carbonate of Potash IUPAC Name: dipotassium carbonate SMILES: [K+].[K+].[O-]C([O-])=O
Potassium Carbonate, Anhydrous, Granular, USP, 99.5-100.5%, Spectrum™ Chemical
CAS: 584-08-7 Molecular Formula: CK2O3 Molecular Weight (g/mol): 138.21 MDL Number: MFCD00011382 InChI Key: BWHMMNNQKKPAPP-UHFFFAOYSA-L IUPAC Name: dipotassium carbonate SMILES: [K+].[K+].[O-]C([O-])=O
- 1
- 2