missing translation for 'onlineSavingsMsg'
Learn More

Potassium Carbonate Anhydrous (Granular Powder/Certified ACS), Fisher Chemical™

Catalog No. P208500
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P208500 500 g Poly Bottle
P2083 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P208500 Supplier Thermo Fisher Scientific Supplier No. P208 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
IUPAC Name dipotassium carbonate
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Melting Point 891°C
Color White
pH 11.5 to 12
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range ≥99.0%
Formula Weight 138.21
Percent Purity ≥99.0%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Density 2.43g/mL
Chemical Name or Material Potassium Carbonate Anhydrous
Show More Show Less