missing translation for 'onlineSavingsMsg'
Learn More

Potassium Carbonate, Anhydrous, Granular, USP, 99.5-100.5%, Spectrum™ Chemical

Catalog No. 18605491
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605491 45 kg Poly Pail
18605489 2.5 kg Poly Pail
18605490 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18605491 Supplier Spectrum Chemical Mfg Cor Supplier No. P123845KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

P1238, 584-08-7, K2CO3

Spectrum™ Chemical Potassium Carbonate, Anhydrous, Granular, USP is used as a drying agent for some non acidic compounds and as a buffering agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
IUPAC Name dipotassium carbonate
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

CAS Max % 1
Physical Form Granular/Lump
Quantity 45 kg
Percent Purity 99.5 to 100.5%
Grade USP
Loss on Drying 0.005
Packaging Poly Pail
Chemical Name or Material Potassium Carbonate, Anhydrous, Granular