missing translation for 'onlineSavingsMsg'
Learn More

Potassium carbonate, 98%, extra pure, anhydrous

Catalog No. AC447620010
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC447620010 1 kg
AC447620050 5 kg
AC447620250 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC447620010 Supplier Thermo Scientific Chemicals Supplier No. 447620010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 584-08-7 | CK2O3 | 138.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
IUPAC Name dipotassium;carbonate
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Melting Point 891.0°C
Quantity 1 kg
Loss on Drying 2.0% max.
Linear Formula K2CO3
Merck Index 15,774
Solubility Information Solubility in water: 1120g/L (20°C). Other solubilities: practically insoluble in ethanol, insoluble in acetone
Formula Weight 138.21
Percent Purity 98%
Grade Extra Pure
Chemical Name or Material Potassium carbonate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
If skin irri
EINECSNumber 209-529-3

RUO – Research Use Only