missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium carbonate, Reagent grade, Solstice
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 2.5 kg |
| Linear Formula | K2CO3 |
| UN Number | NONH for all modes of transport |
| Beilstein | 4267587 |
| IUPAC Name | dipotassium carbonate |
| Formula Weight | 138.21g/mol |
| Percent Purity | ≥98% |
| Grade | Not Available |
| Chemical Name or Material | Potassium carbonate |
Safety and Handling
| Hazard Statement | P261-P305 + P351 + P338 |
| RTECSNumber | TS7750000 |