missing translation for 'onlineSavingsMsg'
Learn More

Potassium Carbonate, ACS Reagent, Reagents

Catalog No. 01337866
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337866 100 g
01337865 2.5 kg
01337864 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337866 Supplier Labchem Inc Supplier No. C1279500100C2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Carbonate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym Dipotassium Carbonate, Carbonate of Potash
IUPAC Name dipotassium carbonate
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Color Colorless/White
pH 11.6
Physical Form Solid
Quantity 100 g
Percent Purity ≥99%
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.29
Chemical Name or Material Potassium Carbonate, ACS Reagent