missing translation for 'onlineSavingsMsg'
Learn More

Potassium Carbonate, Saturated at 20°C, Reagents

Catalog No. 01334202
Change view
Click to view available options
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01334202 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334202 Supplier Labchem Inc Supplier No. CP2027004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Carbonate, Saturated at 20°C, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 584-08-7 , 7732-18-5
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym Dipotassium Carbonate, Carbonate of Potash
IUPAC Name dipotassium carbonate
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Boiling Point 108°C
Color Colorless
pH 11.6
Physical Form Liquid
Quantity 4 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.57
Chemical Name or Material Potassium Carbonate