CAS RN 56-40-6
Glycine (White Crystals or Crystalline Powder), Fisher BioReagents™
CAS: 56-40-6 Molecular Formula: C2H5NO2 Molecular Weight (g/mol): 75.07 MDL Number: MFCD00008131 InChI Key: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC Name: 2-aminoacetic acid SMILES: NCC(O)=O
Glycine (Crystalline/Certified), Fisher Chemical™
CAS: 56-40-6 Molecular Formula: C2H5NO2 Molecular Weight (g/mol): 75.07 MDL Number: MFCD00008131 InChI Key: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC Name: 2-aminoacetic acid SMILES: NCC(O)=O
Glycine (Crystalline Granules or Powder/USP/EP/BP/JP), Fisher Chemical™
CAS: 56-40-6 Molecular Formula: C2H5NO2 Molecular Weight (g/mol): 75.07 MDL Number: MFCD00008131 InChI Key: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC Name: 2-aminoacetic acid SMILES: NCC(O)=O
SDS-PAGE Buffer, 5X Powder, Electrophoresis, Fisher BioReagents
Buffer Concentrate, pH 11.00 (Certified), Fisher Chemical
Glycine (Crystalline Granules or Powder/USP), Fisher Chemical™
CAS: 56-40-6 Molecular Formula: C2H5NO2 Molecular Weight (g/mol): 75.07 MDL Number: MFCD00008131 InChI Key: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC Name: 2-aminoacetic acid SMILES: NCC(O)=O
Thermo Scientific Chemicals Glycine, 0.2M buffer soln., pH 3.0, low endotoxin
CAS: 56-40-6 Molecular Weight (g/mol): 75.067
Thermo Scientific Chemicals Glycine, 99+%, Cell Culture Reagent
CAS: 56-40-6 Molecular Formula: C2H5NO2 Molecular Weight (g/mol): 75.07 MDL Number: MFCD00008131 InChI Key: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC Name: 2-aminoacetic acid SMILES: NCC(O)=O
Thermo Scientific Chemicals Glycine, 99%
CAS: 56-40-6 Molecular Formula: C2H5NO2 Molecular Weight (g/mol): 75.07 MDL Number: MFCD00008131 InChI Key: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC Name: 2-aminoacetic acid SMILES: NCC(O)=O
Glycine, U.S.P. - F.C.C., J.T. Baker™
CAS: 56-40-6 Molecular Formula: C2H5NO2 Molecular Weight (g/mol): 75.07 MDL Number: MFCD00008131 InChI Key: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC Name: 2-aminoacetic acid SMILES: NCC(O)=O
Glycine, Ultrapure Bioreagent, J.T. Baker™
CAS: 56-40-6 Molecular Formula: C2H5NO2 Molecular Weight (g/mol): 75.07 MDL Number: MFCD00008131 InChI Key: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC Name: 2-aminoacetic acid SMILES: NCC(O)=O
Thermo Scientific Chemicals Glycine, ACS, 98.5+%
CAS: 56-40-6 Molecular Formula: C2H5NO2 Molecular Weight (g/mol): 75.07 MDL Number: MFCD00008131 InChI Key: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC Name: 2-aminoacetic acid SMILES: NCC(O)=O
Glycine, Multi-Compendial, U.S.P., J.T. Baker™
CAS: 56-40-6 Molecular Formula: C2H5NO2 Molecular Weight (g/mol): 75.07 MDL Number: MFCD00008131 InChI Key: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC Name: 2-aminoacetic acid SMILES: NCC(O)=O
Glycine, BAKER ANALYZED™ Biochemical Reagent, J.T. Baker™
CAS: 56-40-6 Molecular Formula: C2H5NO2 Molecular Weight (g/mol): 75.07 MDL Number: MFCD00008131 InChI Key: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC Name: 2-aminoacetic acid SMILES: NCC(O)=O
Thermo Scientific Chemicals Glycine, 0.2M buffer soln., pH 3.5, low endotoxin
CAS: 56-40-6 Molecular Weight (g/mol): 75.067