missing translation for 'onlineSavingsMsg'
Learn More

Glycine (Crystalline Granules or Powder/USP), Fisher Chemical™

Catalog No. G48500
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
G48500 500 g Poly Bottle
G48212 2.5 kg Poly Bottle
G4812 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G48500 Supplier Thermo Fisher Scientific Supplier No. FLG48 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point 182°C to 236°C
pH 5.9 to 6.4
Color White
Chloride 0.007% max. (USP)
Hydrolyzable Substance Pass Test (USP)
Ignition Residue 0.1% max. (USP)
Identification Pass Test (USP)
Loss on Drying 0.2% max. (USP)
Packaging Poly Bottle
Linear Formula CH2(NH2)(COOH)
Residual Solvents Meets Requirements (USP)
Sulfate 0.0065% max. (USP)
Density 1.595g/cm³
Quantity 500 g
Formula Weight 75.07
Percent Purity 98.5 to 101.5% (USP)
Grade USP
Physical Form Powder
Chemical Name or Material Glycine
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:1
Flammability:1
Instability:1