missing translation for 'onlineSavingsMsg'
Learn More

SDS-PAGE Buffer, 5X Powder, Electrophoresis, Fisher BioReagents

Catalog No. BP139892
Change view
Click to view available options
Quantity:
92 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP139892 92 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP139892 Supplier Fisher BioReagents Supplier No. BP1398 92
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • Mix of tris-glycine-SDS (TGS) for electrophoresis buffer
  • Tris-glycine-SDS is the most commonly used running buffer for SDS-PAGE
  • Pre-weighed and pre-mixed ingredients to make 1L of 5X tris-glycine-SDS buffer
  • Eliminates the need to weigh ingredients - simply dissolve powder in pure water

Specifications

Chemical Name or Material Tris-Glycine-SDS
CAS 56-40-6, 77-86-1, 151-21-3
Color White
pH 8.3
Physical Form Solid
Quantity 92 g
Packaging Amber glass bottle
Synonym TGS Buffer
Grade Electrophoresis