missing translation for 'onlineSavingsMsg'
Learn More

Glycine, U.S.P. - F.C.C., J.T. Baker™

Catalog No. 02002945
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Wide Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002945 500 g Wide Mouth Amber Glass Bottle
02002944 2 kg Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02002945 Supplier Avantor Supplier No. 058101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56-40-6 | C2H5NO2 | 75.07 g/mol

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Packaging Wide Mouth Amber Glass Bottle
Quantity 500 g
Formula Weight 75.07
Percent Purity ≥98.5%
Grade FCC/USP
Specific Gravity 1L = 1.16kg
Chemical Name or Material Glycine