CAS RN 548-62-9
Crystal Violet (Certified Biological Stain), Fisher Chemical™
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 MDL Number: MFCD00011750 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M Synonym: Gentian Violet,Basic Violet 3 PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Thermo Scientific Chemicals Crystal Violet, pure, indicator
Crystal Violet, C25H30ClN3, CAS Number-548-62-9, 90-94-8, Basic Violet 3, C.I. 42555, Gentian Violet, 100g, 13, 756, CHEBI:41688, 407.99, Pure, Danger, GHS H Statement: Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Causes serious eye damage. May cause cancer. | CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol
Thermo Scientific Chemicals Crystal Violet
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 MDL Number: MFCD00011750 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M Synonym: Basic Violet 3; C.I. 42555 PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Crystal Violet, ACS, 90+%
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 MDL Number: MFCD00011750 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M Synonym: Basic Violet 3; C.I. 42555 PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Thermo Scientific Chemicals Crystal Violet, certified
CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol
Thermo Scientific Chemicals Crystal Violet, pure, high purity biological stain
CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol
Gentian Violet, 2% (w/v), Ricca Chemical
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Crystal Violet Hydrate, TCI America™
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 MDL Number: MFCD00011750 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M Synonym: Basic Violet 3, Hexamethylpararosaniline Chloride PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Gentian (Crystal) Violet, Ricca Chemical
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Crystal Violet Hydrate [for Biochemical Research], TCI America™
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 MDL Number: MFCD00011750 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M Synonym: Basic Violet 3, Hexamethylpararosaniline Chloride PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Gentian Violet, 2% Methanolic, Certified, LabChem™
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Gentian Violet, Reagent Grade, LabChem™
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Gentian Violet, 2% (w/v) in Methanol, Ricca Chemical
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): Mixture InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Gentian Violet, 1% (w/v), Ricca Chemical
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): Mixture InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Crystal Violet Indicator, 1% (w/v) in Glacial Acetic Acid, Ricca Chemical
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): Mixture InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
- 1
- 2