missing translation for 'onlineSavingsMsg'
Learn More

Crystal Violet, ACS, 90+%

Catalog No. AA2286622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2286622 100 g
AA2286614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2286622 Supplier Thermo Scientific Chemicals Supplier No. 02286622

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol

Crystal Violet is used as an acid-base indicator, an alcohol denaturant, a biological stain, a textile dye and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 548-62-9
Molecular Formula C25H30ClN3
Molecular Weight (g/mol) 407.986
MDL Number MFCD00011750
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
Synonym Basic Violet 3; C.I. 42555
PubChem CID 11057
ChEBI CHEBI:41688
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]

Specifications

Melting Point ∼215°C (decomposition)
UN Number UN3077
Beilstein 4077708
Merck Index 14,4395
Solubility Information Soluble in water and chloroform. Slightly soluble in alcohol and glycerol.
Quantity 100 g
Physical Form Powder
Formula Weight 407.99
Percent Purity ≥90%
Grade ACS
Chemical Name or Material Crystal Violet
Show More Show Less
Hazard Category H302-H318-H351
Hazard Statement GHS H Statement
H318-H351-H302
Causes serious eye damage.
Suspected of causing cancer.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P305+P351+P338-P308+P313-P310-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 208-953-6
RTECSNumber BO9000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only