missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Crystal Violet, certified

Catalog No. AC405835000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC405835000 500 g
AC405830250 25 g
AC405831000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC405835000 Supplier Thermo Scientific Chemicals Supplier No. 405835000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Specifications

Chemical Name or Material Crystal Violet
Melting Point 215.0°C
CAS 548-62-9, 90-94-8
Absorption Ratio 0.98 to 1.2 (P-15/P+15)
Dye Content 88% min.
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C25H30ClN3
MDL Number MFCD00011750
Beilstein 13, 756
Merck Index 15, 4430
Color Green
Synonym Basic Violet 3, C.I. 42555, Gentian Violet
Solubility Information Solubility in water: 16g/L (25°C). Other solubilities: soluble in chloroform, 1 g/10 mL alcohol, 1 g/15 mL glycerin, practically insoluble in ether
Quantity 500 g
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
Physical Form Crystalline Powder
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
Molecular Weight (g/mol) 407.986
PubChem CID 11057
ChEBI CHEBI:41688
Formula Weight 407.99
Grade Certified
Show More Show Less

RUO – Research Use Only