missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Crystal Violet, pure, indicator

Catalog No. AC212120250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC212120250 25 g
AC212121000 100 g
AC212125000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC212120250 Supplier Thermo Scientific Chemicals Supplier No. 212120250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Crystal Violet, C25H30ClN3, CAS Number-548-62-9, 90-94-8, Basic Violet 3, C.I. 42555, Gentian Violet, 100g, 13, 756, CHEBI:41688, 407.99, Pure, Danger, GHS H Statement: Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Causes serious eye damage. May cause cancer. | CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Specifications

Chemical Name or Material Crystal Violet
Melting Point 215.0°C
Name Note Indicator, Pure
CAS 548-62-9, 90-94-8
Packaging Glass bottle
Molecular Formula C25H30ClN3
MDL Number MFCD00011750
Beilstein 13, 756
Merck Index 15, 4430
Synonym Basic Violet 3, C.I. 42555, Gentian Violet
Solubility Information Solubility in water: 16g/L (25°C). Other solubilities: soluble in chloroform, 1 g/10 mL alcohol, 1 g/15 mL glycerin, practically insoluble in ether
Quantity 25 g
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
Molecular Weight (g/mol) 407.986
PubChem CID 11057
ChEBI CHEBI:41688
Formula Weight 407.99
Grade Pure
Show More Show Less

RUO – Research Use Only