missing translation for 'onlineSavingsMsg'
Learn More

Gentian (Crystal) Violet, Ricca Chemical

Catalog No. RDCG010025F
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCG010025F 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCG010025F Supplier Ricca Chemical Company Supplier No. RDCG010025F1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 548-62-9
Molecular Formula C25H30ClN3
Molecular Weight (g/mol) 407.986
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
PubChem CID 11057
ChEBI CHEBI:41688
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]

Specifications

CAS Min % 98.0
CAS Max % 100.0
Packaging Pouch
Color Dark Green
pH 2.5 to 3.5
Quantity 25 g
Grade Indicator
Chemical Name or Material Gentian (Crystal)
ShelfLife 60 months