missing translation for 'onlineSavingsMsg'
Learn More

Gentian Violet, Reagent Grade, LabChem™

Catalog No. LC147908
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC147908 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC147908 Supplier LabChem Supplier No. LC147908

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 548-62-9
Molecular Formula C25H30ClN3
Molecular Weight (g/mol) 407.986
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
PubChem CID 11057
ChEBI CHEBI:41688
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]

Specifications

Melting Point 173°C
CAS Max % 100
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1.19g/cm3
Linear Formula C25H30CIN3
Decomposition Information Carbon monoxide; Carbon dioxide; Nitrogen oxides
Density 1.19g/cm3
Color Green
Solubility Information Soluble in water
Quantity 25 g
Physical Form Powder
Formula Weight 407.99
Grade Reagent
Chemical Name or Material Gentian Violet
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Suspected of causing cancer.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not eat, drink or smoke when using this product.
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse month.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 208-953-6
Recommended Storage Room Temperature