missing translation for 'onlineSavingsMsg'
Learn More

Crystal Violet Hydrate [for Biochemical Research], TCI America™

Catalog No. C24605G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C24605G Supplier TCI America Supplier No. C24605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory stain for microbiology and cell culture applications, commonly used in biofilm assays.

Chemical Identifiers

CAS 548-62-9
Molecular Formula C25H30ClN3
Molecular Weight (g/mol) 407.986
MDL Number MFCD00011750
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
Synonym Basic Violet 3, Hexamethylpararosaniline Chloride
PubChem CID 11057
ChEBI CHEBI:41688
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]

Specifications

Melting Point 211°C
UN Number 3077
Color Dark Green
Quantity 5 g
Physical Form Crystal/Powder
Formula Weight 407.99
Grade Laboratory
Chemical Name or Material Crystal Violet Hydrate [for Biochemical Research]
EINECSNumber (4)-0873&(5)-1971
RTECSNumber BO9000000
TSCA Yes