CAS RN 2216-51-5
(-)-Menthol, ≥99% (sum of enantiomers, GC), MilliporeSigma™ Supelco™
(-)-Menthol is a cyclic terpene alcohol that occurs in mentha species.It is responsible for their distinctive odor and flavor.
L-Menthol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Thermo Scientific Chemicals L(-)-Menthol, 99.5%
CAS: 2216-51-5 Molecular Formula: C10H20O Molecular Weight (g/mol): 156.27 MDL Number: MFCD00062979 InChI Key: NOOLISFMXDJSKH-KXUCPTDWSA-N Synonym: l-menthol,--menthol,levomenthol,menthomenthol,l---menthol,hexahydrothymol,menthacamphor,peppermint camphor,1r,2s,5r-2-isopropyl-5-methylcyclohexanol,u.s.p. menthol PubChem CID: 16666 ChEBI: CHEBI:15409 IUPAC Name: (1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol SMILES: CC1CCC(C(C1)O)C(C)C
L-Menthol, 99%
CAS: 2216-51-5 Molecular Formula: C10H20O Molecular Weight (g/mol): 156.269 MDL Number: MFCD00062979 InChI Key: NOOLISFMXDJSKH-KXUCPTDWSA-N Synonym: l-menthol,--menthol,levomenthol,menthomenthol,l---menthol,hexahydrothymol,menthacamphor,peppermint camphor,1r,2s,5r-2-isopropyl-5-methylcyclohexanol,u.s.p. menthol PubChem CID: 16666 ChEBI: CHEBI:15409 IUPAC Name: (1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol SMILES: CC1CCC(C(C1)O)C(C)C
L(-)Menthol, 99%, Ricca Chemical
CAS: 2216-51-5 Molecular Formula: C10H20O Molecular Weight (g/mol): 156.269 InChI Key: NOOLISFMXDJSKH-KXUCPTDWSA-N Synonym: l-menthol,--menthol,levomenthol,menthomenthol,l---menthol,hexahydrothymol,menthacamphor,peppermint camphor,1r,2s,5r-2-isopropyl-5-methylcyclohexanol,u.s.p. menthol PubChem CID: 16666 ChEBI: CHEBI:15409 IUPAC Name: (1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol SMILES: CC1CCC(C(C1)O)C(C)C
(-)-Menthol 99.0+%, TCI America™
CAS: 2216-51-5 Molecular Formula: C10H20O Molecular Weight (g/mol): 156.269 MDL Number: MFCD00062979 InChI Key: NOOLISFMXDJSKH-KXUCPTDWSA-N Synonym: l-menthol,--menthol,levomenthol,menthomenthol,l---menthol,hexahydrothymol,menthacamphor,peppermint camphor,1r,2s,5r-2-isopropyl-5-methylcyclohexanol,u.s.p. menthol PubChem CID: 16666 ChEBI: CHEBI:15409 IUPAC Name: (1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol SMILES: CC1CCC(C(C1)O)C(C)C
Medisca Menthol (L) USP (Crystals) (Natural)
Menthol (L) USP comes as colorless, hexagonal crystals, usually needle-like, or in fused masses, or crystalline powder. Has a pleasant, peppermint-like odor.
Menthol, Crystal, FCC, Spectrum™ Chemical
CAS: 2216-51-5 Molecular Formula: C10H20O Molecular Weight (g/mol): 156.27 InChI Key: NOOLISFMXDJSKH-KXUCPTDWSA-N IUPAC Name: (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol SMILES: CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
L-Menthol, Crystal, USP, 98-102%, Spectrum™ Chemical
CAS: 2216-51-5 Molecular Formula: C10H20O Molecular Weight (g/mol): 156.27 InChI Key: NOOLISFMXDJSKH-KXUCPTDWSA-N IUPAC Name: (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol SMILES: CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O