missing translation for 'onlineSavingsMsg'
Learn More

(-)-Menthol 99.0+%, TCI America™

Catalog No. M0545500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0545500G 500 g
M054525G 25 g
M0545100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M0545500G Supplier TCI America Supplier No. M0545500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2216-51-5
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00062979
InChI Key NOOLISFMXDJSKH-KXUCPTDWSA-N
Synonym l-menthol, --menthol, levomenthol, menthomenthol, l---menthol, hexahydrothymol, menthacamphor, peppermint camphor, 1r,2s,5r-2-isopropyl-5-methylcyclohexanol, u.s.p. menthol
PubChem CID 16666
ChEBI CHEBI:15409
IUPAC Name (1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol
SMILES CC1CCC(C(C1)O)C(C)C

Specifications

Melting Point 43°C
Color White
Boiling Point 216°C
Quantity 500 g
Formula Weight 156.27
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Menthol
EINECSNumber (3)-2333
RTECSNumber OT0700000
TSCA Yes