missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(-)-Menthol, 99.5%

Catalog No. AC125405000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125405000 500 g Glass bottle
AC125400050 5 g Glass bottle
AC125401000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC125405000 Supplier Thermo Scientific Chemicals Supplier No. 125405000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2216-51-5 | C10H20O | 156.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2216-51-5
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00062979
InChI Key NOOLISFMXDJSKH-KXUCPTDWSA-N
Synonym l-menthol, --menthol, levomenthol, menthomenthol, l---menthol, hexahydrothymol, menthacamphor, peppermint camphor, 1r,2s,5r-2-isopropyl-5-methylcyclohexanol, u.s.p. menthol
PubChem CID 16666
ChEBI CHEBI:15409
IUPAC Name (1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol
SMILES CC1CCC(C(C1)O)C(C)C

Specifications

Melting Point 41.0°C to 43.0°C
Color Colorless to White
Density 0.8900g/mL
Boiling Point 212.0°C
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass bottle
Refractive Index 1.46
Quantity 500 g
Beilstein 06,28
Fieser 13,172; 16,203
Merck Index 15,5905
Specific Gravity 0.89
Specific Rotation Condition −51.00 (20.00°C 589nm,c=10,EtOH)
Specific Rotation − 51.00
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in acetic acid,soluble in alcohol,chloroform,ether,,petroleum ether
Formula Weight 156.27
Percent Purity 99.5%
Physical Form Crystals or Crystalline Needles
Chemical Name or Material L(-)-Menthol, 99.7%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
EINECSNumber 218-690-9
RTECSNumber OT0700000
TSCA TSCA

RUO – Research Use Only