missing translation for 'onlineSavingsMsg'
Learn More

L-Menthol, 99%

Catalog No. AAA1047430
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1047430 250 g
AAA1047418 50 g
AAA104740B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1047430 Supplier Thermo Scientific Chemicals Supplier No. A1047430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2216-51-5 | C10H20O | 156.269 g/mol

L-Menthol is used as a cooling agent that strongly activates TRPM8.(Transient Receptor Potential Cation Channel, Subfamily M, Member 8 is a Protein Coding gene). It is used as analgesic (topical), antipruritic agent. It is used as: refreshing agent, food flavor, cool and antipruritic drug, carminative drug. Menthol crystals is used for pers onal care and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in 100% ethanol, methanol (100 mg/ml), ether, and chloroform. Insoluble in water.

Notes
Store at room temperature. Incompatible with Strong oxidizing agents, phenols, potassium permanganate, chromium trioxide, chloral hydrate, thymol, pyrogallol, resorcinol, camphor, butylchloral hydrate, betanaphthol, exalgine.

Chemical Identifiers

CAS 2216-51-5
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00062979
InChI Key NOOLISFMXDJSKH-KXUCPTDWSA-N
Synonym l-menthol, --menthol, levomenthol, menthomenthol, l---menthol, hexahydrothymol, menthacamphor, peppermint camphor, 1r,2s,5r-2-isopropyl-5-methylcyclohexanol, u.s.p. menthol
PubChem CID 16666
ChEBI CHEBI:15409
IUPAC Name (1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol
SMILES CC1CCC(C(C1)O)C(C)C

Specifications

Melting Point 42°C to 45°C
Density 0.89
Boiling Point 212°C to 216°C
Flash Point 93°C (199°F)
Quantity 250 g
Beilstein 1902293
Merck Index 14,5837
Solubility Information Soluble in 100% ethanol,methanol (100mg/ml),ether,and chloroform. Insoluble in water.
Optical Rotation −50° (c=10 in ethanol)
Formula Weight 156.27
Percent Purity 99%
Chemical Name or Material L-Menthol
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 218-690-9
RTECSNumber OT0700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only