missing translation for 'onlineSavingsMsg'
Learn More

L-Menthol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013568
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013568 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013568 Supplier Merck Emd Millipore Supplier No. PHR11161G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 2216-51-5
Flash Point 101°C - closed cup
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Linear Formula C10H20 O
MDL Number MFCD00062979
Synonym (-)-Menthol; (1 R,2 S,5 R)-2-Isopropyl-5-methylcyclohexanol; 5-Methyl-2-(1-methylethyl)cyclohexanol
Type Certified Reference Material
Quantity 1 g
Formula Weight 156.27
Grade Certified Reference Material