missing translation for 'onlineSavingsMsg'
Learn More

Menthol, Crystal, FCC, Spectrum™ Chemical

Catalog No. 18604302
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604302 50 kg Amber Glass Bottle
18604298 25 g Amber Glass Bottle
18604299 125 g Amber Glass Bottle
18604300 500 g Amber Glass Bottle
18604301 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604302 Supplier Spectrum Chemical Mfg Cor Supplier No. M136050KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M1360, 2216-51-5, C10H20O

Spectrum™ Chemical Menthol, Crystal, FCC is used as a flavorant in the food industry. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 2216-51-5
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
InChI Key NOOLISFMXDJSKH-KXUCPTDWSA-N
IUPAC Name (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
SMILES CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O

Specifications

CAS Max % 100%
Melting Range 41-44°C
Packaging Amber Glass Bottle
Quantity 50 kg
Specific Rotation -45° to -51°
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Menthol