missing translation for 'onlineSavingsMsg'
Learn More

Medisca Menthol (L) USP (Crystals) (Natural)

Catalog No. 14185561
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185561 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14185561 Supplier Medisca Supplier No. 052103
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Menthol (L) USP comes as colorless, hexagonal crystals, usually needle-like, or in fused masses, or crystalline powder. Has a pleasant, peppermint-like odor.

Menthol (L) USP comes as colorless, hexagonal crystals, usually needle-like, or in fused masses, or crystalline powder. Has a pleasant, peppermint-like odor.

Specifications

Chemical Name or Material Menthol (L) USP (Crystals) (Natural)
Packaging Pail, 3.5 gallon, White, HDPE, Round
CAS 2216-51-5
Type Drug Standard
Molecular Weight (g/mol) 156.267
Quantity 5 kg
Percent Purity 98 to 102 % (Menthol)
Grade USP